Anti-PPA2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00027068-B01P
103342-538EA 798.46 USD
103342-538
Anti-PPA2 Mouse Polyclonal Antibody
Antibodies
The PPA2 Antibody from Novus Biologicals is a mouse polyclonal antibody to PPA2. This antibody reacts with human. The PPA2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR