Anti-POU4F3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00005459-B01P
103307-812EA 798.46 USD
103307-812
Anti-POU4F3 Mouse Polyclonal Antibody
Antibodies
The POU4F3 Antibody from Novus Biologicals is a mouse polyclonal antibody to POU4F3. This antibody reacts with human. The POU4F3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: POU4F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR