Anti-POU3F2/OCT7 Mouse Monoclonal Antibody [clone: 1H5]

H00005454-M03
103307-808EA 583.08 USD
103307-808
Anti-POU3F2/OCT7 Mouse Monoclonal Antibody [clone: 1H5]
The POU3F2 / OCT7 Antibody (1H5) from Novus Biologicals is a mouse monoclonal antibody to POU3F2 / OCT7. This antibody reacts with human. The POU3F2 / OCT7 Antibody (1H5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POU3F2/OCT7
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR