Anti-POU2F3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83966
103266-534EA 777.05 USD
103266-534
Anti-POU2F3 Rabbit Polyclonal Antibody
Antibodies
The POU2F3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to POU2F3. This antibody reacts with human. The POU2F3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR