Anti-Porimin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56371
103358-948EA 417.23 USD
103358-948
Anti-Porimin Rabbit Polyclonal Antibody
Antibodies
The Porimin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Porimin. This antibody reacts with human, mouse. The Porimin Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Porimin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR