Anti-POR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-16083
102707-236EA 635.51 USD
102707-236
Anti-POR Rabbit Polyclonal Antibody
Antibodies
The POR / Cytochrome P450 Reductase Antibody from Novus Biologicals is a rabbit polyclonal antibody to POR / Cytochrome P450 Reductase. This antibody reacts with human, mouse. The POR / Cytochrome P450 Reductase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR