Anti-POR Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00005447-B01P
103307-790EA 399 USD
103307-790
Anti-POR Mouse Polyclonal Antibody
Antibodies
The POR / Cytochrome P450 Reductase Antibody from Novus Biologicals is a mouse polyclonal antibody to POR / Cytochrome P450 Reductase. This antibody reacts with human. The POR / Cytochrome P450 Reductase Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: POR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR