Anti-PON2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86365
103272-774EA 734.58 USD
103272-774
Anti-PON2 Rabbit Polyclonal Antibody
Antibodies
The PON2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PON2. This antibody reacts with human. The PON2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PON2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR