Anti-PON1 Mouse Polyclonal Antibody

H00005444-A01
103307-780EA 436.92 USD
103307-780
Anti-PON1 Mouse Polyclonal Antibody
The PON1 Antibody from Novus Biologicals is a mouse polyclonal antibody to PON1. This antibody reacts with human. The PON1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PON1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR