Anti-POMT2 Mouse Monoclonal Antibody [clone: 1D9]

Supplier: Novus Biologicals
H00029954-M01
103343-182EA 798.46 USD
103343-182
Anti-POMT2 Mouse Monoclonal Antibody [clone: 1D9]
Antibodies
The POMT2 Antibody (1D9) from Novus Biologicals is a mouse monoclonal antibody to POMT2. This antibody reacts with human. The POMT2 Antibody (1D9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POMT2
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR