Anti-POMT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19891
102710-654EA 635.51 USD
102710-654
Anti-POMT1 Rabbit Polyclonal Antibody
Antibodies
The POMT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to POMT1. This antibody reacts with human. The POMT1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: POMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR