Anti-POLR3D Mouse Monoclonal Antibody [clone: 4E6.]

Supplier: Novus Biologicals
H00000661-M01
103322-738EA 798.46 USD
103322-738
Anti-POLR3D Mouse Monoclonal Antibody [clone: 4E6.]
Antibodies
The POLR3D Antibody (4E6.) from Novus Biologicals is a mouse monoclonal antibody to POLR3D. This antibody reacts with human. The POLR3D Antibody (4E6.) has been validated for the following applications: ELISA.

Type: Primary
Antigen: POLR3D
Clonality: Monoclonal
Clone: 4E6.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR