Anti-POLR2J2 Mouse Monoclonal Antibody [clone: 1B2]

Supplier: Novus Biologicals
H00246721-M01
103347-896EA 798.46 USD
103347-896
Anti-POLR2J2 Mouse Monoclonal Antibody [clone: 1B2]
Antibodies
The POLR2J2 Antibody (1B2) from Novus Biologicals is a mouse monoclonal antibody to POLR2J2. This antibody reacts with human. The POLR2J2 Antibody (1B2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POLR2J2
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR