Anti-POLR2J Mouse Monoclonal Antibody [clone: 1A10]

Supplier: Novus Biologicals
H00005439-M02
103307-762EA 798.46 USD
103307-762
Anti-POLR2J Mouse Monoclonal Antibody [clone: 1A10]
Antibodies
The POLR2J Antibody (1A10) from Novus Biologicals is a mouse monoclonal antibody to POLR2J. This antibody reacts with human. The POLR2J Antibody (1A10) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POLR2J
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR