Anti-POLR2E Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15100
102705-402EA 635.51 USD
102705-402
Anti-POLR2E Rabbit Polyclonal Antibody
Antibodies
The POLR2E Antibody from Novus Biologicals is a rabbit polyclonal antibody to POLR2E. This antibody reacts with human. The POLR2E Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: POLR2E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR