|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | POLR2A |
Antigen Name | RNA Polymerase 2A |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | hsRPB1, hRPB220, EC 2.7.7.48, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II largest subunit, RpIILS, RNA Pol 2, RPB1, MGC75453, RPBh1, RPO2, polymerase (RNA) II (DNA directed) polypeptide A (220kD), POLR2, RNA-directed RNA polymerase II subunit RPB1, polymerase (RNA) II (DNA directed) polypeptide A, RPOL2, RNA polymerase II subunit B1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.6, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, POLRA, 220kDa, RNAPII, RNA Polymerase 2 |
Gene ID | 5430 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IMGILCKKSLGTSAGSLVHISYLEMGHDITRLFYSNIQTVINNWLLIEGHTIGIGDSIADSKTYQDIQNTIKKAKQDVIEVIEKAHNNELEPTPGNTLRQTFENQVNRILNDARDKTGSSAQKSLSEYN |
Purification | Immunogen affinity purified |
Storage Buffer | PBS (pH 7.2) and 40% Glycerol |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |