Anti-POLR1C Mouse Monoclonal Antibody [clone: 2A5-A5]

Supplier: Novus Biologicals
H00009533-M02
103337-180EA 798.46 USD
103337-180
Anti-POLR1C Mouse Monoclonal Antibody [clone: 2A5-A5]
Antibodies
The POLR1C Antibody (2A5-A5) from Novus Biologicals is a mouse monoclonal antibody to POLR1C. This antibody reacts with human. The POLR1C Antibody (2A5-A5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: POLR1C
Clonality: Monoclonal
Clone: 2A5-A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR