Anti-POLR1C Mouse Monoclonal Antibody [clone: M1]

Supplier: Novus Biologicals
H00009533-M05
103337-184EA 798.46 USD
103337-184
Anti-POLR1C Mouse Monoclonal Antibody [clone: M1]
Antibodies
The POLR1C Antibody (M1) from Novus Biologicals is a mouse monoclonal antibody to POLR1C. This antibody reacts with human. The POLR1C Antibody (M1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POLR1C
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR