Anti-POLG2 Mouse Monoclonal Antibody [clone: 1A2]

Supplier: Novus Biologicals
H00011232-M01
103340-426EA 798.46 USD
103340-426
Anti-POLG2 Mouse Monoclonal Antibody [clone: 1A2]
Antibodies
The POLG2 Antibody (1A2) from Novus Biologicals is a mouse monoclonal antibody to POLG2. This antibody reacts with human. The POLG2 Antibody (1A2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POLG2
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR