Anti-POLDIP2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00026073-B01P
103342-080EA 399 USD
103342-080
Anti-POLDIP2 Mouse Polyclonal Antibody
Antibodies
The POLDIP2 Antibody from Novus Biologicals is a mouse polyclonal antibody to POLDIP2. This antibody reacts with human. The POLDIP2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: POLDIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR