Anti-POGZ Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00023126-B01P
103341-018EA 798.46 USD
103341-018
Anti-POGZ Mouse Polyclonal Antibody
Antibodies
The POGZ Antibody from Novus Biologicals is a mouse polyclonal antibody to POGZ. This antibody reacts with human. The POGZ Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: POGZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR