Anti-POF1B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-93938
103285-198EA 734.58 USD
103285-198
Anti-POF1B Rabbit Polyclonal Antibody
Antibodies
The POF1B Antibody from Novus Biologicals is a rabbit polyclonal antibody to POF1B. This antibody reacts with human. The POF1B Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: POF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR