Anti-PNR/NR2E3 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: Novus Biologicals
H00010002-M01
103337-942EA 798.46 USD
103337-942
Anti-PNR/NR2E3 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
The PNR / NR2E3 Antibody (2A12) from Novus Biologicals is a mouse monoclonal antibody to PNR / NR2E3. This antibody reacts with human. The PNR / NR2E3 Antibody (2A12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PNR/NR2E3
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR