Anti-PNPT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00087178-D01P
103345-610EA 429 USD
103345-610
Anti-PNPT1 Rabbit Polyclonal Antibody
Antibodies
The PNPT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PNPT1. This antibody reacts with human, mouse. The PNPT1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PNPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR