|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PNPLA8 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
Antigen Synonyms | EC 3.1.1.5, Intracellular membrane-associated calcium-independent phospholipase A2 gamma, calcium-independent phospholipase A2-gamma, membrane-associated calcium-independent phospholipase A2 gamma, iPLA2 gamma, IPLA2GIPLA2(GAMMA), Patatin-like phospholipase domain-containing protein 8, PNPLA-gamma, iPLA2-gamma, patatin-like phospholipase domain containing 8, IPLA22, IPLA2-2 |
Gene ID | 50640 |
Concentration | 0.84 mg/ml |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PNPLA8. The exact sequence is proprietary. |
Purification | Immunogen affinity purified |
Storage Buffer | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |