Anti-PNPLA6 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46109
103409-000EA 564.74 USD
103409-000
Anti-PNPLA6 Goat Polyclonal Antibody
Antibodies
The PNPLA6 Antibody from Novus Biologicals is a goat polyclonal antibody to PNPLA6. This antibody reacts with human, mouse. The PNPLA6 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: PNPLA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR