Anti-PNPLA2 Goat Polyclonal Antibody

NBP1-00225
103408-962EA 466.15 USD
103408-962
Anti-PNPLA2 Goat Polyclonal Antibody
The PEDF R / PNPLA2 / ATGL Antibody from Novus Biologicals is a goat polyclonal antibody to PEDF R / PNPLA2 / ATGL. This antibody reacts with human. The PEDF R / PNPLA2 / ATGL Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: PNPLA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR