Anti-PNP Mouse Monoclonal Antibody [clone: 6E5.]

Supplier: Novus Biologicals
H00004860-M01
103331-872EA 798.46 USD
103331-872
Anti-PNP Mouse Monoclonal Antibody [clone: 6E5.]
Antibodies
The Purine Nucleoside Phosphorylase / PNP Antibody (6E5.) from Novus Biologicals is a mouse monoclonal antibody to Purine Nucleoside Phosphorylase / PNP. This antibody reacts with human. The Purine Nucleoside Phosphorylase / PNP Antibody (6E5.) has been validated for the following applications: Western Blot, ELISA.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PNP
Clonality: Monoclonal
Clone: 6E5.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR