Anti-PNMA2 Mouse Monoclonal Antibody [clone: 2D4]

H00010687-M03
103339-444EA 583.08 USD
103339-444
Anti-PNMA2 Mouse Monoclonal Antibody [clone: 2D4]
The PNMA2 Antibody (2D4) from Novus Biologicals is a mouse monoclonal antibody to PNMA2. This antibody reacts with human. The PNMA2 Antibody (2D4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PNMA2
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR