Anti-PNMA1 Mouse Monoclonal Antibody [clone: 1C3]

Supplier: Novus Biologicals
H00009240-M02
103336-496EA 798.46 USD
103336-496
Anti-PNMA1 Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
The PNMA1 Antibody (1C3) from Novus Biologicals is a mouse monoclonal antibody to PNMA1. This antibody reacts with human. The PNMA1 Antibody (1C3) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PNMA1
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR