Anti-PNMA1 Mouse Monoclonal Antibody [clone: 4G6]

Supplier: Novus Biologicals
H00009240-M01
103336-494EA 798.46 USD
103336-494
Anti-PNMA1 Mouse Monoclonal Antibody [clone: 4G6]
Antibodies
The PNMA1 Antibody (4G6) from Novus Biologicals is a mouse monoclonal antibody to PNMA1. This antibody reacts with human, mouse. The PNMA1 Antibody (4G6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PNMA1
Clonality: Monoclonal
Clone: 4G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR