Anti-PNK Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87257
103270-520EA 777.05 USD
103270-520
Anti-PNK Rabbit Polyclonal Antibody
Antibodies
The PNK Antibody from Novus Biologicals is a rabbit polyclonal antibody to PNK. This antibody reacts with human, mouse. The PNK Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR