Anti-PMS2 Mouse Monoclonal Antibody [clone: 4G4]

H00005395-M01
103307-690EA 583.08 USD
103307-690
Anti-PMS2 Mouse Monoclonal Antibody [clone: 4G4]
The PMS2 Antibody (4G4) from Novus Biologicals is a mouse monoclonal antibody to PMS2. This antibody reacts with human. The PMS2 Antibody (4G4) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PMS2
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR