Anti-PMP34 Mouse Monoclonal Antibody [clone: 3B10]

Supplier: Novus Biologicals
H00010478-M04
103338-990EA 399 USD
103338-990
Anti-PMP34 Mouse Monoclonal Antibody [clone: 3B10]
Antibodies
The PMP34 Antibody (3B10) from Novus Biologicals is a mouse monoclonal antibody to PMP34. This antibody reacts with human. The PMP34 Antibody (3B10) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PMP34
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR