Anti-PMFBP1 Rabbit Polyclonal Antibody

NBP1-86487
103272-840EA 607.69 USD
103272-840
Anti-PMFBP1 Rabbit Polyclonal Antibody
The PMFBP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PMFBP1. This antibody reacts with human. The PMFBP1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PMFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR