Anti-PMCA1 Mouse Monoclonal Antibody [clone: 3E2.]

Supplier: Novus Biologicals
H00000490-M01
103322-370EA 798.46 USD
103322-370
Anti-PMCA1 Mouse Monoclonal Antibody [clone: 3E2.]
Antibodies
The PMCA1 Antibody (3E2.) from Novus Biologicals is a mouse monoclonal antibody to PMCA1. This antibody reacts with human. The PMCA1 Antibody (3E2.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PMCA1
Clonality: Monoclonal
Clone: 3E2.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR