Anti-PLZF Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19870
102710-612EA 663.82 USD
102710-612
Anti-PLZF Rabbit Polyclonal Antibody
Antibodies
The PLZF Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLZF. This antibody reacts with human. The PLZF Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PLZF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR