Anti-PLXNA4 Rabbit Polyclonal Antibody

NBP2-19866
102710-604EA 493.85 USD
102710-604
Anti-PLXNA4 Rabbit Polyclonal Antibody
The Plexin A4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Plexin A4. This antibody reacts with human. The Plexin A4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLXNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR