Anti-PLXNA2 Mouse Monoclonal Antibody [clone: 2G5]

Supplier: Novus Biologicals
H00005362-M06
103307-670EA 798.46 USD
103307-670
Anti-PLXNA2 Mouse Monoclonal Antibody [clone: 2G5]
Antibodies
The Plexin A2 Antibody (2G5) from Novus Biologicals is a mouse monoclonal antibody to Plexin A2. This antibody reacts with human, mouse, rat. The Plexin A2 Antibody (2G5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PLXNA2
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR