Anti-PLSCR4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86165
103269-240EA 734.58 USD
103269-240
Anti-PLSCR4 Rabbit Polyclonal Antibody
Antibodies
The PLSCR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLSCR4. This antibody reacts with human. The PLSCR4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLSCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR