Anti-PLSCR1 Mouse Monoclonal Antibody [clone: 3B4]

Supplier: Novus Biologicals
H00005359-M02
103307-662EA 798.46 USD
103307-662
Anti-PLSCR1 Mouse Monoclonal Antibody [clone: 3B4]
Antibodies
The Phospholipid Scramblase 1 / PLSCR1 Antibody (3B4) from Novus Biologicals is a mouse monoclonal antibody to Phospholipid Scramblase 1 / PLSCR1. This antibody reacts with human. The Phospholipid Scramblase 1 / PLSCR1 Antibody (3B4) has been validated for the following applications: ELISA, Immunoprecipitation.

Type: Primary
Antigen: PLSCR1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR