Anti-PLS1 Mouse Monoclonal Antibody [clone: 3G10]

Supplier: Novus Biologicals
H00005357-M04
103307-652EA 798.46 USD
103307-652
Anti-PLS1 Mouse Monoclonal Antibody [clone: 3G10]
Antibodies
The PLS1 Antibody (3G10) from Novus Biologicals is a mouse monoclonal antibody to PLS1. This antibody reacts with human. The PLS1 Antibody (3G10) has been validated for the following applications: Western Blot, ELISA, Knockout Validated.

Type: Primary
Antigen: PLS1
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR