Anti-PLRP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005407-D01P
103307-706EA 844.62 USD
103307-706
Anti-PLRP1 Rabbit Polyclonal Antibody
Antibodies
The PLRP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLRP1. This antibody reacts with human. The PLRP1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR