Anti-PLK4 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00010733-B01P
103339-500EA 399 USD
103339-500
Anti-PLK4 Mouse Polyclonal Antibody
Antibodies
The PLK4 Antibody from Novus Biologicals is a mouse polyclonal antibody to PLK4. This antibody reacts with human. The PLK4 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR