Anti-PLK3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00001263-D01P
103324-122EA 844.62 USD
103324-122
Anti-PLK3 Rabbit Polyclonal Antibody
Antibodies
The PLK3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLK3. This antibody reacts with human. The PLK3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR