Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLK1 Mouse Monoclonal Antibody [clone: AZ44]

Supplier: Novus Biologicals
NB100-2747
The PLK1 Antibody (AZ44) from Novus Biologicals is a mouse monoclonal antibody to PLK1. This antibody reacts with xenopus. The PLK1 Antibody (AZ44) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: AZ44
Conjugation: Unconjugated
Epitope: This antibody is thought to bind to a phosphorylated epitope. However, this has not yet been proved, but it does stain in a mitotic specific manner.
Host: Mouse
Isotype: IgG1
Reactivity: Xenopus




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB100-2747 103357-494 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PLK1
Reactivity Xenopus
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone AZ44
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
Antigen Synonyms polo like kinase, PLK-1, EC 2.7.11.21, polo (Drosophia)-like kinase, PLK, serine/threonine-protein kinase PLK1, Serine/threonine-protein kinase 13, cell cycle regulated protein kinase, polo-like kinase 1polo-like kinase (Drosophila), STPK13, EC 2.7.11
Gene ID 5347
Epitope This antibody is thought to bind to a phosphorylated epitope. However, this has not yet been proved, but it does stain in a mitotic specific manner.
Concentration 1.0 mg/ml
Immunogen Baculo expressed Polo-like-kinase from Xenopus laevis
Purification Protein A purified
Storage Buffer PBS
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty