Anti-PLK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56651
103360-912EA 491.08 USD
103360-912
Anti-PLK1 Rabbit Polyclonal Antibody
Antibodies
The PLK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLK1. This antibody reacts with human, mouse, rat, porcine, bovine, canine, primate. The PLK1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR