Anti-PLGF Rabbit Polyclonal Antibody

H00005228-D01P
103332-684EA 583.08 USD
103332-684
Anti-PLGF Rabbit Polyclonal Antibody
The PLGF Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLGF. This antibody reacts with human. The PLGF Antibody has been validated for the following applications: Western Blot, Proximity Ligation Assay.

Type: Primary
Antigen: PLGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR