Anti-PLEKHO2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84101
103266-608EA 734.58 USD
103266-608
Anti-PLEKHO2 Rabbit Polyclonal Antibody
Antibodies
The PLEKHO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLEKHO2. This antibody reacts with human. The PLEKHO2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLEKHO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR