Anti-PLEKHM1P Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00440456-B01P
103348-534EA 798.46 USD
103348-534
Anti-PLEKHM1P Mouse Polyclonal Antibody
Antibodies
The PLEKHM1P Antibody from Novus Biologicals is a mouse polyclonal antibody to PLEKHM1P. This antibody reacts with human. The PLEKHM1P Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PLEKHM1P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR